siRNA Details
| Gene Symbol: 1700008I05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027952.3 | NP_082228.2 | 8 | 1029 | ||
| GenBank mRNA | AK005772.1 | - | 8 | 1030 | ||
| BC141320.1 | - | 8 | 1010 | 9055932 | ||
| BC141322.1 | - | 8 | 1010 | 9055934 | ||

1700008I05Rik
Genomic Map