siRNA Details
| Gene Symbol: Fam178b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126046.1 | NP_001119518.1 | 13 | 1791 | ||
| NM_027957.1 | NP_082233.1 | 4 | 414 | |||
| GenBank mRNA | AK006300.1 | - | 4 | 415 | ||
| BC114576.1 | - | 4 | 333 | |||
| BU605266.1 | - | 1 | 40 | |||

Fam178b
Genomic Map