siRNA Details
| Gene Symbol: Fbxo30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168297.1 | NP_001161769.1 | 2 | 1790 | ||
| NM_027968.3 | NP_082244.2 | 2 | 1824 | |||
| GenBank mRNA | AK006369.1 | - | 2 | 1791 | ||
| AK083479.1 | - | 2 | 1781 | |||
| AK135274.1 | - | 2 | 1783 | |||
| BC037691.1 | - | 1 | 703 | 3601336 | ||

Fbxo30
Genomic Map