siRNA Details
Gene Symbol: 1700023E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027970.1 | NP_082246.1 | 6 | 1006 | ||
GenBank mRNA | AK006270.1 | - | 6 | 986 | ||
BC053422.1 | - | 6 | 1006 | 6741654 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700023E05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027970.1 | NP_082246.1 | 6 | 1006 | ||
GenBank mRNA | AK006270.1 | - | 6 | 986 | ||
BC053422.1 | - | 6 | 1006 | 6741654 |