siRNA Details
| Gene Symbol: Cenpu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027973.3 | NP_082249.1 | 11, 12 | 974 | ||
| GenBank mRNA | AK006479.1 | - | 11, 12 | 983 | ||
| AK050386.1 | - | 11, 12 | 975 | |||
| AK081703.1 | - | 11, 12 | 962 | |||
| AY366362.1 | - | 7, 8 | 506 | |||
| BC094675.1 | - | 12, 13 | 1094 | |||
| BC117784.2 | - | 11, 12 | 975 | 40092112 | ||
| BC117785.2 | - | 11, 12 | 975 | 40092113 | ||

Cenpu
Genomic Map