siRNA Details
| Gene Symbol: 2310003H01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027980.2 | NP_082256.2 | 9 | 2632 | ||
| GenBank mRNA | AK082976.1 | - | 9 | 2636 | ||
| AK147900.1 | - | 2724 | ||||
| AK148234.1 | - | 9 | 2619 | |||
| BC034203.1 | - | 9 | 2503 | 4986832 | ||
| BC048376.1 | - | 5 | 971 | |||

2310003H01Rik
Genomic Map