siRNA Details
| Gene Symbol: 2310002L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027104.3 | NP_081380.2 | 2 | 411 | ||
| NM_027981.1 | NP_082257.1 | 2 | 411 | |||
| GenBank mRNA | AK009097.1 | - | 2 | 412 | ||
| AK009124.1 | - | 2 | 411 | |||
| BC115483.1 | - | 2 | 355 | 40100595 | ||
| BC115484.1 | - | 2 | 355 | 40100599 | ||

2310002L09Rik
Genomic Map