siRNA Details
| Gene Symbol: CMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018024.2 | NP_001018024.1 | 3 | 906 | ||
| GenBank mRNA | BC002600.1 | - | 3 | 868 | 3161408 | |
| X64644.1 | - | 3 | 892 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CMC4| Gene Symbol: CMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018024.2 | NP_001018024.1 | 3 | 906 | ||
| GenBank mRNA | BC002600.1 | - | 3 | 868 | 3161408 | |
| X64644.1 | - | 3 | 892 | |||