siRNA Details
| Gene Symbol: Haus5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027999.1 | NP_082275.1 | 15 | 1398 | ||
| GenBank mRNA | AK009476.1 | - | 15 | 1378 | ||
| AK129228.1 | - | 11 | 2666 | |||
| AK153920.1 | - | 15 | 1380 | |||
| BC023723.1 | - | 15 | 1348 | |||
| BC048724.1 | - | 1 | 23 | |||
| BC089002.1 | - | 15 | 1367 | |||
| BC118934.1 | - | 15 | 1373 | 40090953 | ||
| BC118935.1 | - | 15 | 1373 | 40090954 | ||

Haus5
Genomic Map