siRNA Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293703.1 | NP_001280632.1 | 4 | 319 | ||
| NM_028000.1 | NP_082276.1 | 4 | 319 | |||
| GenBank mRNA | AA184457.1 | - | 4 | 319 | ||
| AK007542.1 | - | 2 | 280 | |||
| AK009455.1 | - | 4 | 286 | |||
| AK144593.1 | - | 4 | 304 | |||
| BB857717.1 | - | 4 | 320 | |||
| BC023111.1 | - | 4 | 266 | |||
| BC039626.1 | - | 3 | 134 | |||
| BC062173.1 | - | 4 | 252 | |||
| BC089556.1 | - | 4 | 263 | |||
| BC099489.1 | - | 4 | 270 | |||

Ppapdc1b
Genomic Map