siRNA Details
| Gene Symbol: Epgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053087.2 | NP_444317.1 | 3 | 186 | ||
| GenBank mRNA | AJ291391.1 | - | 3 | 186 | ||
| AK010136.1 | - | 2 | 134 | |||
| BC119291.1 | - | 3 | 239 | 8734040 | ||
| BC119293.1 | - | 3 | 239 | 8734042 | ||
| BC145041.1 | - | 3 | 286 | |||

Epgn
Genomic Map