siRNA Details
| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028011.2 | NP_082287.1 | 8, 9 | 907 | ||
| GenBank mRNA | AF395837.1 | - | 8, 9 | 871 | ||
| AJ505007.1 | - | 8, 9 | 868 | |||
| AK009826.1 | - | 6, 7 | 642 | |||
| AK033712.1 | - | 8, 9 | 909 | |||
| AK083630.1 | - | 869 | ||||
| BC004710.1 | - | 1, 2 | 1264 | 3499918 | ||
| BC020011.1 | - | 2, 3 | 1540 | |||

Tom1l1
Genomic Map