siRNA Details
| Gene Symbol: 2310067B10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028014.3 | NP_082290.2 | 31 | 4216 | ||
| GenBank mRNA | AK010082.1 | - | 2 | 215 | ||
| AK047622.1 | - | 17 | 2350 | |||
| BC007157.1 | - | 18 | 2462 | |||
| BC030865.1 | - | 16 | 1983 | |||
| BC053088.1 | - | 31 | 4131 | 5709104 | ||

2310067B10Rik
Genomic Map