siRNA Details
| Gene Symbol: Cers5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028015.2 | NP_082291.1 | 9 | 1124 | ||
| GenBank mRNA | AK010241.1 | - | 9 | 1052 | ||
| AK075694.1 | - | 9 | 1044 | |||
| AK145589.1 | - | 9 | 1032 | |||
| AK145762.1 | - | 9 | 1046 | |||
| AK146220.1 | - | 9 | 1046 | |||
| AK146417.1 | - | 9 | 1016 | |||
| AY029533.1 | - | 9 | 1044 | |||
| BC010670.1 | - | 3 | 345 | |||
| BC043059.1 | - | 1071 | 6419704 | |||
| BC046797.1 | - | 9 | 1124 | 6403129 | ||

Cers5
Genomic Map