siRNA Details
| Gene Symbol: 2410016O06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023633.3 | NP_076122.2 | 1 | 1697 | ||
| GenBank mRNA | AB041553.1 | - | 1 | 1636 | ||
| AK010521.1 | - | 1 | 803 | |||
| AK080870.1 | - | 1 | 1643 | |||
| AK213032.1 | - | 1 | 284 | |||
| BC052152.2 | - | 1 | 1611 | |||
| BC059809.1 | - | 1 | 1648 | |||
| BC138094.1 | - | 1 | 1697 | 8861113 | ||

2410016O06Rik
Genomic Map