siRNA Details
| Gene Symbol: Myh14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271538.1 | NP_001258467.1 | 32 | 4580 | ||
| NM_001271540.1 | NP_001258469.1 | 33 | 4522 | |||
| NM_028021.3 | NP_082297.1 | 31 | 4457 | |||
| GenBank mRNA | AY205605.1 | - | 31 | 4457 | ||
| AY363100.1 | - | 33 | 4522 | |||
| BC041796.1 | - | 6 | 818 | |||
| BC054465.1 | - | 2 | 286 | |||
| EF602040.1 | - | 32 | 4580 | |||

Myh14
Genomic Map