siRNA Details
| Gene Symbol: Nkiras2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028024.2 | NP_082300.1 | 2, 3 | 170 | ||
| GenBank mRNA | AK010371.1 | - | 2, 3 | 170 | ||
| AK015918.1 | - | 175 | ||||
| AK052645.1 | - | 2, 3 | 170 | |||
| AK083475.1 | - | 2, 3 | 213 | |||
| AK149696.1 | - | 2, 3 | 314 | |||
| AK169878.1 | - | 2, 3 | 195 | |||
| BC013469.1 | - | 1, 2 | 123 | 4219892 | ||
| BC024398.1 | - | 1, 2 | 113 | 5003001 | ||

Nkiras2
Genomic Map