siRNA Details
| Gene Symbol: MTHFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005957.4 | NP_005948.3 | 10 | 1839 | ||
| XM_005263458.3 | XP_005263515.1 | 10 | 2676 | |||
| XM_005263460.4 | XP_005263517.1 | 10 | 2294 | |||
| XM_005263462.4 | XP_005263519.1 | 10 | 1727 | |||
| XM_005263463.3 | XP_005263520.1 | 10 | 1856 | |||
| XM_011541495.2 | XP_011539797.1 | 10 | 2673 | |||
| XM_011541496.2 | XP_011539798.1 | 10 | 2676 | |||
| XM_017001328.1 | XP_016856817.1 | 10 | 2676 | |||
| GenBank mRNA | AB209113.1 | - | 9 | 1877 | ||
| AJ237672.1 | - | 9 | 1739 | |||
| AK312907.1 | - | 10 | 1700 | |||
| BC053509.1 | - | 10 | 1661 | 6374885 | ||
| JQ429447.1 | - | 6 | 1115 | |||

MTHFR
Genomic Map