siRNA Details
| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028036.3 | NP_082312.3 | 7 | 888 | ||
| GenBank mRNA | AK010487.1 | - | 7 | 813 | ||
| AK046207.1 | - | 7 | 926 | |||
| BC005637.1 | - | 7 | 890 | 3156734 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco6| Gene Symbol: Tmco6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028036.3 | NP_082312.3 | 7 | 888 | ||
| GenBank mRNA | AK010487.1 | - | 7 | 813 | ||
| AK046207.1 | - | 7 | 926 | |||
| BC005637.1 | - | 7 | 890 | 3156734 | ||