siRNA Details
| Gene Symbol: Acad10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028037.4 | NP_082313.2 | 20 | 3212 | ||
| GenBank mRNA | AK010568.1 | - | 10 | 1663 | ||
| AK050332.1 | - | 20 | 3416 | |||
| BC027825.1 | - | 20 | 3188 | 5343012 | ||
| BC029047.1 | - | 16 | 2361 | |||
 Back To Top
 Back To Top
 Acad10
Acad10 Genomic Map
Genomic Map Back To Top
 Back To Top