siRNA Details
| Gene Symbol: Ddx54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028041.2 | NP_082317.1 | 13 | 1565 | ||
| GenBank mRNA | AF319547.1 | - | 13 | 1925 | ||
| AK010484.1 | - | 10 | 1145 | |||
| AK028274.1 | - | 13 | 1579 | |||
| AK031534.1 | - | 13 | 1575 | |||
| AK031679.1 | - | 13 | 1583 | |||
| AK147715.1 | - | 13 | 1580 | |||
| AK152989.1 | - | 13 | 1581 | |||
| AK155138.1 | - | 13 | 1577 | |||
| AK167603.1 | - | 13 | 1579 | |||
| AK168455.1 | - | 13 | 1579 | |||
| AK169255.1 | - | 13 | 1578 | |||
| AK170967.1 | - | 13 | 1577 | |||
| AK184886.1 | - | 1 | 29 | |||
| BC021900.1 | - | 4 | 570 | |||
| BC043699.1 | - | 13 | 1565 | 5344158 | ||
| BC055026.1 | - | 9 | 1008 | |||
| BC062920.1 | - | 13 | 1568 | |||
| BC071251.1 | - | 7 | 862 | |||

Ddx54
Genomic Map