siRNA Details
| Gene Symbol: 1600014K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028046.1 | NP_082322.1 | 2 | 475 | ||
| GenBank mRNA | AK005459.1 | - | 1 | 433 | ||
| BC049727.1 | - | 2 | 466 | 6770312 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1600014K23Rik| Gene Symbol: 1600014K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028046.1 | NP_082322.1 | 2 | 475 | ||
| GenBank mRNA | AK005459.1 | - | 1 | 433 | ||
| BC049727.1 | - | 2 | 466 | 6770312 | ||