siRNA Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028047.2 | NP_082323.1 | 7 | 1066 | ||
| GenBank mRNA | AK005126.1 | - | 7 | 1066 | ||
| AK039368.1 | - | 7 | 1065 | |||
| AK141576.1 | - | 7 | 1057 | |||
| AK169651.1 | - | 7 | 990 | |||
| BC034211.1 | - | 7 | 981 | 4988361 | ||

Smg9
Genomic Map