siRNA Details
| Gene Symbol: Smg9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028047.2 | NP_082323.1 | 6 | 875 | ||
| GenBank mRNA | AK005126.1 | - | 6 | 875 | ||
| AK039368.1 | - | 6 | 874 | |||
| AK141576.1 | - | 6 | 866 | |||
| AK161454.1 | - | 6 | 885 | |||
| AK169651.1 | - | 6 | 799 | |||
| BC034211.1 | - | 6 | 790 | 4988361 | ||

Smg9
Genomic Map