siRNA Details
| Gene Symbol: Lmntd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028050.2 | NP_082326.1 | 7 | 800 | ||
| GenBank mRNA | BC119093.1 | - | 7 | 804 | 8733842 | |
| BC119119.1 | - | 7 | 804 | 8733868 | ||
| BC144834.1 | - | 7 | 834 | 9053382 | ||

Lmntd2
Genomic Map