siRNA Details
| Gene Symbol: Fndc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173182.2 | NP_775274.2 | 22, 23 | 2918 | ||
| GenBank mRNA | AB098596.1 | - | 22, 23 | 2856 | ||
| AK136013.1 | - | 1, 2 | 116 | |||
| AK147346.1 | - | 23, 24 | 2928 | |||
| AK220297.1 | - | 22, 23 | 2918 | |||
| AY234860.1 | - | 4, 5 | 660 | |||
| BC067389.1 | - | 22, 23 | 2924 | 30431862 | ||
| DQ192037.1 | - | 21, 22 | 2824 | |||

Fndc3b
Genomic Map