siRNA Details
| Gene Symbol: Fndc3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173182.2 | NP_775274.2 | 3 | 266 | ||
| GenBank mRNA | AB098596.1 | - | 3 | 204 | ||
| AK045099.1 | - | 3 | 361 | |||
| AK147346.1 | - | 4 | 277 | |||
| AK220297.1 | - | 3 | 266 | |||
| BC067389.1 | - | 3 | 272 | 30431862 | ||
| DQ192037.1 | - | 2 | 172 | |||

Fndc3b
Genomic Map