siRNA Details
| Gene Symbol: Btbd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028055.4 | NP_082331.1 | 3 | 1345 | ||
| GenBank mRNA | AK005160.1 | - | 3 | 1351 | ||
| AK032100.1 | - | 3 | 1347 | |||
| AK046050.1 | - | 3 | 1346 | |||
| BC057599.1 | - | 2 | 2622 | |||
| BC060053.1 | - | 2 | 2653 | |||
| BC122876.1 | - | 3 | 1328 | 40107633 | ||

Btbd17
Genomic Map