siRNA Details
| Gene Symbol: Btbd17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028055.4 | NP_082331.1 | 2 | 321 | ||
| GenBank mRNA | AK005160.1 | - | 2 | 327 | ||
| AK032100.1 | - | 2 | 323 | |||
| AK046050.1 | - | 2 | 322 | |||
| BC057599.1 | - | 2 | 271 | |||
| BC060053.1 | - | 2 | 302 | |||
| BC122876.1 | - | 2 | 304 | 40107633 | ||

Btbd17
Genomic Map