siRNA Details
Gene Symbol: Cyb5r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028057.2 | NP_082333.1 | 5 | 440 | ||
GenBank mRNA | AK005159.1 | - | 5 | 441 | ||
AK049717.1 | - | 4 | 662 | |||
AK159663.1 | - | 5 | 410 | |||
AK167178.1 | - | 5 | 434 | |||
BC016266.1 | - | 5 | 649 | 4924006 | ||
BC024618.1 | - | 5 | 418 | 4235599 | ||
JX420288.1 | - | 520 | ||||
JX420290.1 | - | 5 | 617 |
