siRNA Details
| Gene Symbol: Fundc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028058.3 | NP_082334.1 | 2 | 164 | ||
| GenBank mRNA | AK005163.1 | - | 2 | 165 | ||
| AK007686.1 | - | 1 | 201 | |||
| AK168868.1 | - | 2 | 151 | |||
| BC012515.1 | - | 2 | 106 | 2651125 | ||
| BC030487.1 | - | 2 | 170 | 5374103 | ||

Fundc1
Genomic Map