siRNA Details
| Gene Symbol: Fundc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028058.3 | NP_082334.1 | 2 | 250 | ||
| GenBank mRNA | AK005163.1 | - | 2 | 251 | ||
| AK007686.1 | - | 1 | 287 | |||
| AK168868.1 | - | 2 | 237 | |||
| AK210161.1 | - | 11 | ||||
| BC012515.1 | - | 2 | 192 | 2651125 | ||
| BC030487.1 | - | 2 | 256 | 5374103 | ||

Fundc1
Genomic Map