siRNA Details
| Gene Symbol: Zfp654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028059.2 | NP_082335.1 | 1 | 1703 | ||
| GenBank mRNA | AK005508.1 | - | 1 | 1756 | ||
| BC027760.1 | - | 1 | 1405 | |||
| BC068018.1 | - | 1 | 1703 | 6336013 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp654| Gene Symbol: Zfp654 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028059.2 | NP_082335.1 | 1 | 1703 | ||
| GenBank mRNA | AK005508.1 | - | 1 | 1756 | ||
| BC027760.1 | - | 1 | 1405 | |||
| BC068018.1 | - | 1 | 1703 | 6336013 | ||