siRNA Details
| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081320.2 | NP_001074789.2 | 3 | 557 | ||
| NM_028061.1 | NP_082337.1 | 3 | 424 | |||
| GenBank mRNA | AK005419.1 | - | 2 | 624 | ||
| AK087683.1 | - | 3 | 505 | |||
| AK164441.1 | - | 3 | 431 | |||
| BC043107.1 | - | 3 | 1043 | |||
| BC060107.1 | - | 3 | 399 | |||
| BC137590.1 | - | 3 | 534 | 8860609 | ||

Cyb561d1
Genomic Map