siRNA Details
| Gene Symbol: Cyb561d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081320.2 | NP_001074789.2 | 3 | 1217 | ||
| NM_028061.1 | NP_082337.1 | 3 | 1084 | |||
| GenBank mRNA | AK005419.1 | - | 2 | 1284 | ||
| AK087683.1 | - | 3 | 1168 | |||
| AK164441.1 | - | 3 | 1091 | |||
| BC043107.1 | - | 3 | 1703 | |||
| BC060107.1 | - | 3 | 1059 | |||
| BC137590.1 | - | 3 | 1194 | 8860609 | ||

Cyb561d1
Genomic Map