siRNA Details
| Gene Symbol: Slc39a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028064.2 | NP_082340.1 | 5 | 1083 | ||
| GenBank mRNA | AK005535.1 | - | 5 | 1123 | ||
| AK146977.1 | - | 5 | 1124 | |||
| AK147107.1 | - | 5 | 1102 | |||
| AK154089.1 | - | 5 | 1602 | |||
| AK155467.1 | - | 5 | 1602 | |||
| BC023498.1 | - | 5 | 1083 | 5011088 | ||

Slc39a4
Genomic Map