siRNA Details
Gene Symbol: MTNR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005959.3 | NP_005950.1 | 2 | 1033 | ||
XM_011542839.2 | XP_011541141.1 | 2 | 930 | |||
XM_017017777.1 | XP_016873266.1 | 2 | 1336 | |||
GenBank mRNA | AY521019.1 | - | 2 | 930 | ||
BC069163.1 | - | 2 | 1033 | 7216982 | ||
U25341.1 | - | 2 | 942 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
