siRNA Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023645.3 | NP_076134.1 | 5, 6 | 1127 | ||
| GenBank mRNA | AJ404004.1 | - | 5, 6 | 889 | ||
| AK007835.1 | - | 5, 6 | 1120 | |||
| AK035899.1 | - | 5, 6 | 1128 | |||
| AK053315.1 | - | 5, 6 | 1120 | |||
| AK079701.1 | - | 5, 6 | 1120 | |||
| AK170300.1 | - | 5, 6 | 1156 | |||
| AK198736.1 | - | 2, 3 | 233 | |||

Kdelc1
Genomic Map