siRNA Details
| Gene Symbol: 1810055G02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028077.2 | NP_082353.1 | 3, 4 | 500 | ||
| GenBank mRNA | AK007868.1 | - | 3, 4 | 501 | ||
| AK154828.1 | - | 3, 4 | 822 | |||
| BC019471.1 | - | 3, 4 | 494 | 4161022 | ||

1810055G02Rik
Genomic Map