siRNA Details
| Gene Symbol: Ush1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163733.1 | NP_001157205.1 | 24 | 2454 | ||
| NM_001291182.1 | NP_001278111.1 | 24 | 2454 | |||
| NM_023649.2 | NP_076138.2 | 19 | 1542 | |||
| NM_153677.2 | NP_710143.2 | 24 | 2454 | |||
| GenBank mRNA | AF228924.1 | - | 19 | 1542 | ||
| AF228925.1 | - | 24 | 2454 | |||
| AK008274.2 | - | 3 | 211 | |||
| AY103465.1 | - | 24 | 2454 | |||
| AY246556.1 | - | 24 | 2428 | |||
| BC010819.1 | - | 19 | 1544 | 4217086 | ||
| BY708209.1 | - | 3 | 208 | |||

Ush1c
Genomic Map