siRNA Details
| Gene Symbol: Ugt2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028094.3 | NP_082370.2 | 1 | 296 | ||
| GenBank mRNA | AK008601.1 | - | 1 | 278 | ||
| AK050327.1 | - | 1 | 297 | |||
| BC025795.1 | - | 1 | 272 | 5133743 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a3| Gene Symbol: Ugt2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028094.3 | NP_082370.2 | 1 | 296 | ||
| GenBank mRNA | AK008601.1 | - | 1 | 278 | ||
| AK050327.1 | - | 1 | 297 | |||
| BC025795.1 | - | 1 | 272 | 5133743 | ||