siRNA Details
| Gene Symbol: Mettl10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028095.1 | NP_082371.1 | 4 | 420 | ||
| GenBank mRNA | AK008476.1 | - | 4 | 405 | ||
| BC038323.1 | - | 4 | 375 | |||
| BC096622.1 | - | 4 | 404 | 30445095 | ||
| BC116375.2 | - | 4 | 418 | 40101137 | ||

Mettl10
Genomic Map