siRNA Details
| Gene Symbol: 2010300C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028096.1 | NP_082372.1 | 4, 5 | 626 | ||
| GenBank mRNA | BC072639.1 | - | 4, 5 | 626 | 30355303 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

2010300C02Rik| Gene Symbol: 2010300C02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028096.1 | NP_082372.1 | 4, 5 | 626 | ||
| GenBank mRNA | BC072639.1 | - | 4, 5 | 626 | 30355303 | |