siRNA Details
| Gene Symbol: Dusp11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028099.4 | NP_082375.4 | 7, 8 | 755 | ||
| GenBank mRNA | AK008492.1 | - | 2, 3 | 139 | ||
| AK081629.1 | - | 7, 8 | 732 | |||
| AK088941.1 | - | 7, 8 | 752 | |||
| BC028640.1 | - | 3, 4 | 182 | |||
| BC067028.1 | - | 7, 8 | 709 | 30364073 | ||

Dusp11
Genomic Map