siRNA Details
| Gene Symbol: Jmjd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028101.4 | NP_082377.2 | 8 | 731 | ||
| GenBank mRNA | AK011308.1 | - | 8 | 728 | ||
| AK148079.1 | - | 8 | 709 | |||
| AK155810.1 | - | 8 | 733 | |||
| AK172073.1 | - | 8 | 728 | |||
| BC010800.1 | - | 8 | 693 | 4235072 | ||

Jmjd8
Genomic Map