siRNA Details
| Gene Symbol: Tpx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001141975.1 | NP_001135447.1 | 12 | 1606 | ||
| NM_001141976.1 | NP_001135448.1 | 12 | 1622 | |||
| NM_001141977.1 | NP_001135449.1 | 12 | 1810 | |||
| NM_001141978.1 | NP_001135450.1 | 12 | 1609 | |||
| NM_028109.4 | NP_082385.3 | 12 | 1797 | |||
| GenBank mRNA | AK011311.2 | - | 9 | 1165 | ||
| AK090101.1 | - | 12 | 1560 | |||
| AK153957.1 | - | 12 | 1544 | |||
| AK167030.1 | - | 9 | 1246 | |||
| BC060619.1 | - | 12 | 1698 | 6812536 | ||

Tpx2
Genomic Map