siRNA Details
| Gene Symbol: Dennd2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001093754.2 | NP_001087223.1 | 11 | 1480 | ||
| NM_001278941.1 | NP_001265870.1 | 12 | 1559 | |||
| NM_028110.2 | NP_082386.2 | 11 | 1478 | |||
| GenBank mRNA | AK008546.1 | - | 11 | 1284 | ||
| AK050033.1 | - | 12 | 1560 | |||
| AK157279.1 | - | 11 | 1481 | |||
| BC008266.1 | - | 11 | 1468 | 2811014 | ||
| BC043727.1 | - | 11 | 1266 | 5035764 | ||

Dennd2d
Genomic Map