siRNA Details
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153780.3 | NP_722475.1 | 2, 3 | 266 | ||
| GenBank mRNA | AK011776.1 | - | 2, 3 | 243 | ||
| AK029464.1 | - | 2, 3 | 229 | |||
| AK138138.1 | - | 2, 3 | 267 | |||
| BC023890.1 | - | 2, 3 | 225 | 5252244 | ||

2610044O15Rik8
Genomic Map