siRNA Details
| Gene Symbol: Gm1976 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082664.1 | - | 3 | 181 | ||
| AK137318.1 | - | 3 | 182 | |||
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153780.3 | NP_722475.1 | 4 | 342 | ||
| GenBank mRNA | AK011776.1 | - | 4 | 319 | ||
| AK029464.1 | - | 4 | 305 | |||
| AK138138.1 | - | 4 | 343 | |||
| BC023890.1 | - | 4 | 301 | 5252244 | ||

Gm1976
Genomic Map