siRNA Details
| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153780.3 | NP_722475.1 | 5 | 1054 | ||
| GenBank mRNA | AK011776.1 | - | 5 | 1031 | ||
| AK138138.1 | - | 5 | 1055 | |||
| BC023890.1 | - | 5 | 1013 | 5252244 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610044O15Rik8| Gene Symbol: 2610044O15Rik8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153780.3 | NP_722475.1 | 5 | 1054 | ||
| GenBank mRNA | AK011776.1 | - | 5 | 1031 | ||
| AK138138.1 | - | 5 | 1055 | |||
| BC023890.1 | - | 5 | 1013 | 5252244 | ||